accessibility menu, dialog, popup

UserName

H3COON

CAS RN 39931-77-6

IUPAC Name: ethyl 2-(pyridin-3-yl)acetate
Synonyms: ethyl 2-(pyridin-3-yl)acetate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: RPWXYCRIAGBAGY-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CN=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)