missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Piperidinecarboxylate 98.0+%, TCI America™
$32.72 - $115.51
Chemical Identifiers
| CAS | 5006-62-2 |
|---|---|
| Molecular Formula | C8H15NO2 |
| Molecular Weight (g/mol) | 157.213 |
| MDL Number | MFCD00005991 |
| InChI Key | XIWBSOUNZWSFKU-UHFFFAOYSA-N |
| Synonym | ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate |
| PubChem CID | 98969 |
| IUPAC Name | ethyl piperidine-3-carboxylate |
| SMILES | CCOC(=O)C1CCCNC1 |
Chemical Identifiers
| 5006-62-2 | |
| 157.213 | |
| XIWBSOUNZWSFKU-UHFFFAOYSA-N | |
| 98969 | |
| CCOC(=O)C1CCCNC1 |
| C8H15NO2 | |
| MFCD00005991 | |
| ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate | |
| ethyl piperidine-3-carboxylate |
Specifications
| 5006-62-2 | |
| 104°C | |
| MFCD00005991 | |
| ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate | |
| CCOC(=O)C1CCCNC1 | |
| 157.213 | |
| 157.21 | |
| Liquid |
| Yellow | |
| C8H15NO2 | |
| 5 g | |
| XIWBSOUNZWSFKU-UHFFFAOYSA-N | |
| ethyl piperidine-3-carboxylate | |
| 98969 | |
| ≥98.0% (GC,T) | |
| Ethyl 3-Piperidinecarboxylate |
Safety and Handling
TSCA : No