missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Piperidinecarboxylate 98.0+%, TCI America™

Catalog No. E05845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05845G 5 g
E058425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05845G Supplier TCI America Supplier No. E05845G

Chemical Identifiers

CAS 5006-62-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00005991
InChI Key XIWBSOUNZWSFKU-UHFFFAOYSA-N
Synonym ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
PubChem CID 98969
IUPAC Name ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Piperidinecarboxylate