Learn More
Ethyl 3-phenylpropionate, 98+%
CAS: 2021-28-5 | C11H14O2 | 178.231 g/mol
Supplier: Thermo Scientific Chemicals A1305530
Description
Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.
Solubility
Miscible with oils and ethanol. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| Ethyl 3-phenylpropionate | |
| 1.016 | |
| 107°C (224°F) | |
| 1.494 | |
| 250 g | |
| ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate | |
| JAGZUIGGHGTFHO-UHFFFAOYSA-N | |
| ethyl 3-phenylpropanoate | |
| 16237 | |
| ≥98% |
| 2021-28-5 | |
| 118°C (3 mmHg) | |
| C11H14O2 | |
| MFCD00009206 | |
| 1910432 | |
| Miscible with oils and ethanol. Immiscible with water. | |
| CCOC(=O)CCC1=CC=CC=C1 | |
| 178.231 | |
| 178.23 |
Chemical Identifiers
| 2021-28-5 | |
| 178.231 | |
| JAGZUIGGHGTFHO-UHFFFAOYSA-N | |
| 16237 | |
| CCOC(=O)CCC1=CC=CC=C1 |
| C11H14O2 | |
| MFCD00009206 | |
| ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate | |
| ethyl 3-phenylpropanoate |
Safety and Handling
EINECSNumber : 217-966-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only