missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenylpropionate, 98+%

Catalog No. p-7020931
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1305518 50 g
AAA1305530 250 g
AAA130550B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1305518 Supplier Thermo Scientific Chemicals Supplier No. A1305518

CAS: 2021-28-5 | C11H14O2 | 178.231 g/mol

Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

Solubility
Miscible with oils and ethanol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2021-28-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009206
InChI Key JAGZUIGGHGTFHO-UHFFFAOYSA-N
Synonym ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate
PubChem CID 16237
IUPAC Name ethyl 3-phenylpropanoate
SMILES CCOC(=O)CCC1=CC=CC=C1

Specifications

Density 1.016
Boiling Point 118°C (3 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.494
Quantity 50 g
Beilstein 1910432
Solubility Information Miscible with oils and ethanol. Immiscible with water.
Formula Weight 178.23
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-phenylpropionate
EINECSNumber 217-966-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only