missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxo-3-(2-thienyl)propionate 98.0+%, TCI America™

Supplier:  TCI America E10731G

 View more versions of this product

Catalog No. E10731G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-Oxo-3-(2-thienyl)propionate
130°C
MFCD00542649
3-Oxo-3-(2-thienyl)propionic Acid Ethyl Ester
CCOC(=O)CC(=O)C1=CC=CS1
198.236
198.24
Liquid
13669-10-8
C9H10O3S
1 g
VKSDKUXHVLZDHO-UHFFFAOYSA-N
ethyl 3-oxo-3-thiophen-2-ylpropanoate
255159
≥98.0% (GC)

Chemical Identifiers

13669-10-8
198.236
VKSDKUXHVLZDHO-UHFFFAOYSA-N
255159
CCOC(=O)CC(=O)C1=CC=CS1
C9H10O3S
MFCD00542649
3-Oxo-3-(2-thienyl)propionic Acid Ethyl Ester
ethyl 3-oxo-3-thiophen-2-ylpropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions