accessibility menu, dialog, popup

UserName

H3COOOS

CAS RN 13669-10-8

IUPAC Name: ethyl 3-oxo-3-(thiophen-2-yl)propanoate
Synonyms: ethyl 3-oxo-3-(thiophen-2-yl)propanoate
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: VKSDKUXHVLZDHO-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results