missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Methylthio)propionate 98.0+%, TCI America™

Supplier:  TCI America E079325G

Catalog No. E079325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-(Methylthio)propionate
Yellow
C6H12O2S
25 g
YSNWHRKJEKWJNY-UHFFFAOYSA-N
ethyl 3-methylsulfanylpropanoate
61592
148.22
Liquid
13327-56-5
192°C
MFCD00039900
ethyl 3-methylthio propionate, ethyl 3-methylthio propanoate, ethyl 3-methylsulfanyl propanoate, propanoic acid, 3-methylthio-, ethyl ester, ethyl methyl mercaptopropionate, ethyl 3-methylmercaptopropionate, 3-methylthio propionic acid ethylester, 3-methylthio propanoic acid ethyl ester, unii-1avt374nii, ethyl methylthiopropanoate
CCOC(=O)CCSC
148.22
CHEBI:87503
≥98.0% (GC)

Chemical Identifiers

13327-56-5
148.22
YSNWHRKJEKWJNY-UHFFFAOYSA-N
61592
ethyl 3-methylsulfanylpropanoate
C6H12O2S
MFCD00039900
ethyl 3-methylthio propionate, ethyl 3-methylthio propanoate, ethyl 3-methylsulfanyl propanoate, propanoic acid, 3-methylthio-, ethyl ester, ethyl methyl mercaptopropionate, ethyl 3-methylmercaptopropionate, 3-methylthio propionic acid ethylester, 3-methylthio propanoic acid ethyl ester, unii-1avt374nii, ethyl methylthiopropanoate
CHEBI:87503
CCOC(=O)CCSC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1395

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions