missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Methylpyrazole-5-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 80°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | ethyl 3-methyl-1h-pyrazole-5-carboxylate, ethyl 3-methylpyrazole-5-carboxylate, 5-methyl-1h-pyrazole-3-carboxylic acid ethyl ester, 3-methyl-1h-pyrazole-5-carboxylic acid ethyl ester, 1h-pyrazole-3-carboxylic acid, 5-methyl-, ethyl ester, ethyl3-methylpyrazole-5-carboxylate, 5-methyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl3-methyl-1h-pyrazole-5-carboxylate, ethyl 5-methyl-2h-pyrazole-3-carboxylate, pubchem7843 |
| Formula Weight | 154.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 3-Methylpyrazole-5-carboxylate |
Safety and Handling
| TSCA | No |