missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Methylindole-2-carboxylate 98.0+%, TCI America™

Catalog No. E10905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10905G 5 g
E10901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10905G Supplier TCI America Supplier No. E10905G
Only null left

Chemical Identifiers

CAS 26304-51-8
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00225365
InChI Key OZQXZSIVKVCSDF-UHFFFAOYSA-N
Synonym 3-Methylindole-2-carboxylic Acid Ethyl Ester
PubChem CID 593963
IUPAC Name ethyl 3-methyl-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC=C2N1)C

Specifications

Melting Point 137°C
Color Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Methylindole-2-carboxylate