accessibility menu, dialog, popup

UserName

H3COOHNH3C

CAS RN 26304-51-8

IUPAC Name: ethyl 3-methyl-1H-indole-2-carboxylate
Synonyms: ethyl 3-methyl-1H-indole-2-carboxylate
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: OZQXZSIVKVCSDF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results