missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%

CAS: 22232-54-8 | C7H10N2O2S | 186.229 g/mol

$420.14 - $420.14

Chemical Identifiers

CAS 22232-54-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD00027421
InChI Key CFOYWRHIYXMDOT-UHFFFAOYSA-N
Synonym carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
PubChem CID 31072
ChEBI CHEBI:617099
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C1=S)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0821814
View Documents
Thermo Scientific Chemicals
L0821814
25 g
Each for $420.14
Only null left
 
Description

Description

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

Solubility
Soluble in water, ethanol, chloroform and acetone. Sightly soluble in water.

Notes
Light sensitive. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

22232-54-8
186.229
CFOYWRHIYXMDOT-UHFFFAOYSA-N
31072
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
C7H10N2O2S
MFCD00027421
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CHEBI:617099
CCOC(=O)N1C=CN(C1=S)C

Specifications

22232-54-8
5.0 to 7.0
MFCD00027421
144339
14,1798
Soluble in water,ethanol,chloroform and acetone. Sightly soluble in water.
CCOC(=O)N1C=CN(C1=S)C
186.229
CHEBI:617099
97%
122°C to 126°C
C7H10N2O2S
25 g
Light sensitive
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CFOYWRHIYXMDOT-UHFFFAOYSA-N
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
31072
186.24
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P281-P308+P313-P501c

H360

EINECSNumber : 244-854-4

RTECSNumber : NJ2441000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only