missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methoxyphenylacetate, 98%

Catalog No. AAA1507006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1507006 5 g
AAA1507014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1507006 Supplier Thermo Scientific Chemicals Supplier No. A1507006

CAS: 35553-92-5 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35553-92-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026897
InChI Key XXVVNHCWPHMLEZ-UHFFFAOYSA-N
Synonym ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
PubChem CID 2774971
IUPAC Name ethyl 2-(3-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=CC=C1)OC

Specifications

Boiling Point 109°C to 110°C (1 mmHg)
Quantity 5 g
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 3-methoxyphenylacetate
EINECSNumber 252-614-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only