missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-Methoxybenzoyl)acetate 98.0+%, TCI America™

Catalog No. E11215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11215G 5 g
E11211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11215G Supplier TCI America Supplier No. E11215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27834-99-7
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00086704
InChI Key FDPPVAYPZOORBP-UHFFFAOYSA-N
Synonym (3-Methoxybenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(3-Methoxyphenyl)-3-oxopropionate, 3-(3-Methoxyphenyl)-3-oxopropionic Acid Ethyl Ester
PubChem CID 2759696
IUPAC Name ethyl 3-(3-methoxyphenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(OC)=CC=C1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (3-Methoxybenzoyl)acetate
EINECSNumber (3)-1745
TSCA No