accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 27834-99-7

IUPAC Name: ethyl 3-(3-methoxyphenyl)-3-oxopropanoate
Synonyms: ethyl 3-(3-methoxyphenyl)-3-oxopropanoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: FDPPVAYPZOORBP-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC(OC)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results