missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Iodobenzoate 98.0+%, TCI America™

Catalog No. I05365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05365G 5 g
I053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05365G Supplier TCI America Supplier No. I05365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58313-23-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00044769
InChI Key POGCXCWRMMXDAQ-UHFFFAOYSA-N
Synonym ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709
PubChem CID 143542
IUPAC Name ethyl 3-iodobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)I

Specifications

Color Yellow
Boiling Point 161°C
Quantity 5 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Iodobenzoate