missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Indoleacetate 98.0+%, TCI America™
Supplier: TCI America E087825G
Specifications
| Ethyl 3-Indoleacetate | |
| 44°C | |
| C12H13NO2 | |
| 25 g | |
| HUDBDWIQSIGUDI-UHFFFAOYSA-N | |
| ethyl 2-(1H-indol-3-yl)acetate | |
| 13067 | |
| ≥98.0% (GC) |
| 778-82-5 | |
| 166°C | |
| MFCD00005635 | |
| ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate | |
| CCOC(=O)CC1=CNC2=CC=CC=C21 | |
| 203.241 | |
| 203.24 | |
| Crystalline Powder |
Chemical Identifiers
| 778-82-5 | |
| 203.241 | |
| HUDBDWIQSIGUDI-UHFFFAOYSA-N | |
| 13067 | |
| CCOC(=O)CC1=CNC2=CC=CC=C21 |
| C12H13NO2 | |
| MFCD00005635 | |
| ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate | |
| ethyl 2-(1H-indol-3-yl)acetate |
Safety and Handling
RTECSNumber : NL3528000
TSCA : No