missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Indoleacetate 98.0+%, TCI America™

Catalog No. p-7129197
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08785G 5 g
E087825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08785G Supplier TCI America Supplier No. E08785G
Only null left

Chemical Identifiers

CAS 778-82-5
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005635
InChI Key HUDBDWIQSIGUDI-UHFFFAOYSA-N
Synonym ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate
PubChem CID 13067
IUPAC Name ethyl 2-(1H-indol-3-yl)acetate
SMILES CCOC(=O)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 44°C
Boiling Point 166°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Indoleacetate
RTECSNumber NL3528000
TSCA No