missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Hydroxyhexanoate 98.0+%, TCI America™
$126.76 - $339.25
Chemical Identifiers
| CAS | 2305-25-1 |
|---|---|
| Molecular Formula | C8H16O3 |
| Molecular Weight (g/mol) | 160.213 |
| MDL Number | MFCD00036604 |
| InChI Key | LYRIITRHDCNUHV-UHFFFAOYSA-N |
| Synonym | hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate |
| PubChem CID | 61293 |
| ChEBI | CHEBI:23997 |
| IUPAC Name | ethyl 3-hydroxyhexanoate |
| SMILES | CCCC(CC(=O)OCC)O |
Chemical Identifiers
| 2305-25-1 | |
| 160.213 | |
| LYRIITRHDCNUHV-UHFFFAOYSA-N | |
| 61293 | |
| ethyl 3-hydroxyhexanoate |
| C8H16O3 | |
| MFCD00036604 | |
| hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate | |
| CHEBI:23997 | |
| CCCC(CC(=O)OCC)O |
Specifications
| 2305-25-1 | |
| 92°C | |
| MFCD00036604 | |
| hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate | |
| CCCC(CC(=O)OCC)O | |
| 160.213 | |
| CHEBI:23997 | |
| ≥98.0% (GC) | |
| Ethyl 3-Hydroxyhexanoate |
| Colorless | |
| C8H16O3 | |
| 25 g | |
| LYRIITRHDCNUHV-UHFFFAOYSA-N | |
| ethyl 3-hydroxyhexanoate | |
| 61293 | |
| 160.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1430
TSCA : Yes