missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Hydroxyhexanoate 98.0+%, TCI America™
Supplier: TCI America E0788100G
Specifications
| Ethyl 3-Hydroxyhexanoate | |
| Colorless | |
| C8H16O3 | |
| 100 g | |
| LYRIITRHDCNUHV-UHFFFAOYSA-N | |
| ethyl 3-hydroxyhexanoate | |
| 61293 | |
| 160.21 | |
| Liquid |
| 2305-25-1 | |
| 92°C | |
| MFCD00036604 | |
| hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate | |
| CCCC(CC(=O)OCC)O | |
| 160.213 | |
| CHEBI:23997 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2305-25-1 | |
| 160.213 | |
| LYRIITRHDCNUHV-UHFFFAOYSA-N | |
| 61293 | |
| ethyl 3-hydroxyhexanoate |
| C8H16O3 | |
| MFCD00036604 | |
| hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate | |
| CHEBI:23997 | |
| CCCC(CC(=O)OCC)O |
Safety and Handling
EINECSNumber : (2)-1430
TSCA : Yes