missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxybutyrate, 99%

CAS: 5405-41-4 | C6H12O3 | 132.16 g/mol

$59.91 - $150.10

Chemical Identifiers

CAS 5405-41-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004545
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
PubChem CID 62572
ChEBI CHEBI:87685
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118540250
SDS View Documents
Thermo Scientific Chemicals
118540250
25 g Glass bottle
Each for $59.91
Only null left
 
AC118541000
SDS View Documents
Thermo Scientific Chemicals
118541000
100 g Glass bottle
Each for $150.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5405-41-4
132.16
OMSUIQOIVADKIM-UHFFFAOYNA-N
CHEBI:87685
CCOC(=O)CC(C)O
C6H12O3
MFCD00004545
62572
ethyl 3-hydroxybutanoate

Specifications

5405-41-4
1.0100g/mL
64°C
98.5% min. (GC)
C6H12O3
CH3CH(OH)CH2CO2C2H5
25 g
14,162
ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
OMSUIQOIVADKIM-UHFFFAOYNA-N
ethyl 3-hydroxybutanoate
62572
132.16
Liquid
Colorless
170°C
Authentic
Glass bottle
1.4195 to 1.4215
MFCD00004545
03, 309
1.01
Solubility in water: soluble
CCOC(=O)CC(C)O
132.16
CHEBI:87685
99%
Ethyl 3-hydroxybutyrate, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-456-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only