missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxybutyrate, 98+%

Catalog No. AAB2152114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2152114 25 g
AAB2152122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2152114 Supplier Thermo Scientific Chemicals Supplier No. B2152114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5405-41-4 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5405-41-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004545
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
PubChem CID 62572
ChEBI CHEBI:87685
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Density 1.013
Boiling Point 169°C to 171°C
Flash Point 64°C (148°F)
Refractive Index 1.421
Linear Formula CH3CHOHCH2CO2CH2CH3
Quantity 25 g
Beilstein 1446190
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-hydroxybutyrate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 226-456-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only