missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Hydroxy-2-naphthoate 98.0+%, TCI America™

Catalog No. E12491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12491G 1 g
E12495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12491G Supplier TCI America Supplier No. E12491G

Chemical Identifiers

CAS 7163-25-9
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.236
MDL Number MFCD00220608
InChI Key CVEOWBRXZJEZRQ-UHFFFAOYSA-N
Synonym 3-Hydroxy-2-naphthoic Acid Ethyl Ester
PubChem CID 737040
IUPAC Name ethyl 3-hydroxynaphthalene-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2C=C1O

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 291°C
Quantity 1 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Hydroxy-2-naphthoate