missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-fluorobenzoate, 98+%

Catalog No. AAL0798409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0798409 10 g
AAL0798418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0798409 Supplier Thermo Scientific Chemicals Supplier No. L0798409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 451-02-5 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 451-02-5
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00000336
InChI Key SMMIKBXLEWTSJD-UHFFFAOYSA-N
Synonym ethyl m-fluorobenzoate, benzoic acid, 3-fluoro-, ethyl ester, 3-fluorobenzoic acid ethyl ester, 3-fluoro-benzoic acid ethyl ester, benzoic acid, m-fluoro-, ethyl ester, pubchem3470, 3-fluorobenzoic acid ethyl, acmc-209k0o, ethyl3-fluorobenzoate, 3-fluoro-benzoicaciethylester
PubChem CID 67975
IUPAC Name ethyl 3-fluorobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)F

Specifications

Density 1.136
Boiling Point 208°C
Flash Point 82°C (179°F)
Odor Recognizable
Refractive Index 1.485
Quantity 10 g
Beilstein 2045876
Formula Weight 168.17
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-fluorobenzoate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-191-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only