missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyclopropyl-3-oxopropionate, 96%

Catalog No. p-7032336
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3375603 1 g
AAH3375606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3375603 Supplier Thermo Scientific Chemicals Supplier No. H3375603

CAS: 24922-02-9 | C8H12O3 | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24922-02-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD00184570
InChI Key LFSVADABIDBSBV-UHFFFAOYSA-N
Synonym 3-cyclopropyl-3-oxo-propionic acid ethyl ester, ethyl 3-cyclopropyl-3-oxo-propanoate, ethyl 3-cyclopropyl-3-oxopropionate, ethyl 3-cyclopropyl-3-oxo-propionate, ethyl beta-oxo-cyclopropane propionate, ksc205o7f, ethyl cyclopropylcarbonylacetate, carbethoxymethyl cyclopropyl ketone, ethyl ?-oxo-cyclopropanepropionate, ethyl3-cyclopropyl-3-oxopropanoate
PubChem CID 262979
IUPAC Name ethyl 3-cyclopropyl-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1CC1

Specifications

Boiling Point 73°C (1 mmHg)
Quantity 1 g
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material Ethyl 3-cyclopropyl-3-oxopropionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only