missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Cyclohexene-1-carboxylate 95.0+%, TCI America™

Catalog No. C09665ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09665ML 5 mL
C096625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09665ML Supplier TCI America Supplier No. C09665ML

Chemical Identifiers

CAS 15111-56-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00037158
InChI Key BYNLHLLOIZDKIC-UHFFFAOYSA-N
Synonym 3-Cyclohexene-1-carboxylic Acid Ethyl Ester
PubChem CID 85803
IUPAC Name ethyl cyclohex-3-ene-1-carboxylate
SMILES CCOC(=O)C1CCC=CC1

Specifications

Color Yellow
Boiling Point 195°C
Quantity 5 mL
Formula Weight 154.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Cyclohexene-1-carboxylate