accessibility menu, dialog, popup

UserName

H3COO

CAS RN 15111-56-5

IUPAC Name: ethyl cyclohex-3-ene-1-carboxylate
Synonyms: ethylcyclohexene-1-carboxylate
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: BYNLHLLOIZDKIC-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCC=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results