missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(Chloromethyl)benzoate 96.0+%, TCI America™
Supplier: TCI America E066125G
Specifications
| Ethyl 3-(Chloromethyl)benzoate | |
| 169°C | |
| 25 g | |
| 3-(Chloromethyl)benzoic Acid Ethyl Ester | |
| CCOC(=O)C1=CC(=CC=C1)CCl | |
| 198.646 | |
| 198.65 | |
| Liquid |
| 54589-54-7 | |
| C10H11ClO2 | |
| 3265 | |
| ZJNVMXXFCNKXLT-UHFFFAOYSA-N | |
| ethyl 3-(chloromethyl)benzoate | |
| 14812190 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 54589-54-7 | |
| 198.646 | |
| 3-(Chloromethyl)benzoic Acid Ethyl Ester | |
| ethyl 3-(chloromethyl)benzoate |
| C10H11ClO2 | |
| ZJNVMXXFCNKXLT-UHFFFAOYSA-N | |
| 14812190 | |
| CCOC(=O)C1=CC(=CC=C1)CCl |
Safety and Handling
TSCA : No