missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Bromobenzoate 98.0+%, TCI America™

Catalog No. p-7119278
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18085G 5 g
B180825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18085G Supplier TCI America Supplier No. B18085G
Only null left

Chemical Identifiers

CAS 24398-88-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00013529
InChI Key QAUASTLEZAPQTB-UHFFFAOYSA-N
Synonym ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
PubChem CID 90488
IUPAC Name ethyl 3-bromobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Br

Specifications

Boiling Point 261°C
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Bromobenzoate