missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(Benzylamino)propionate 95.0+%, TCI America™
$48.46 - $870.51
Chemical Identifiers
| CAS | 23583-21-3 |
|---|---|
| Molecular Formula | C12H17NO2 |
| Molecular Weight (g/mol) | 207.273 |
| MDL Number | MFCD00085541 |
| InChI Key | HCTJHQFFNDLDPF-UHFFFAOYSA-N |
| Synonym | ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate |
| PubChem CID | 90196 |
| IUPAC Name | ethyl 3-(benzylamino)propanoate |
| SMILES | CCOC(=O)CCNCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B157925ML
|
TCI America
B157925ML |
25 mL |
Each for $48.46
|
|
|||||
|
B1579500ML
|
TCI America
B1579500ML |
500 mL |
Each for $870.51
|
|
|||||
Chemical Identifiers
| 23583-21-3 | |
| 207.273 | |
| HCTJHQFFNDLDPF-UHFFFAOYSA-N | |
| 90196 | |
| CCOC(=O)CCNCC1=CC=CC=C1 |
| C12H17NO2 | |
| MFCD00085541 | |
| ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate | |
| ethyl 3-(benzylamino)propanoate |
Specifications
| 23583-21-3 | |
| Yellow | |
| MFCD00085541 | |
| HCTJHQFFNDLDPF-UHFFFAOYSA-N | |
| ethyl 3-(benzylamino)propanoate | |
| 207.273 | |
| 207.27 | |
| Liquid |
| 135°C | |
| C12H17NO2 | |
| ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate | |
| CCOC(=O)CCNCC1=CC=CC=C1 | |
| 25 mL | |
| 90196 | |
| ≥95.0% (GC) | |
| Ethyl 3-(Benzylamino)propionate |
Safety and Handling
EINECSNumber : (3)-4218
TSCA : No
Recommended Storage : Refrigerator