accessibility menu, dialog, popup

UserName

CAS RN 23583-21-3

H3COONH

CAS RN 23583-21-3

IUPAC Name: ethyl 3-(benzylamino)propanoate
Synonyms: ethyl 3-(benzylamino)propanoate
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: HCTJHQFFNDLDPF-UHFFFAOYSA-N
SMILES: CCOC(=O)CCNCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)