missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Benzylamino)-3-oxopropionate 98.0+%, TCI America™

Catalog No. E10985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10985G 5 g
E10981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10985G Supplier TCI America Supplier No. E10985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29689-63-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD28128094
InChI Key MPAGGJGTAABULX-UHFFFAOYSA-N
Synonym 3-(Benzylamino)-3-oxopropionic Acid Ethyl Ester, Ethyl (N-Benzylcarbamoyl)acetate, (N-Benzylcarbamoyl)acetic Acid Ethyl Ester, Ethyl N-Benzylmalonamate, N-Benzylmalonamic Acid Ethyl Ester
PubChem CID 11379032
IUPAC Name ethyl 2-(benzylcarbamoyl)acetate
SMILES CCOC(=O)CC(=O)NCC1=CC=CC=C1

Specifications

Melting Point 51°C
Color Yellow
Quantity 5 g
Formula Weight 221.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-(Benzylamino)-3-oxopropionate