CAS RN 29689-63-2
IUPAC Name:
ethyl 2-(benzylcarbamoyl)acetate
Synonyms:
ethyl 2-(benzylcarbamoyl)acetate
Molecular Weight (g/mol):
221.26
Molecular Formula:
C12H15NO3
InChi Key:
MPAGGJGTAABULX-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)NCC1=CC=CC=C1
1
–
1
of
1
results
1
Ethyl 3-(Benzylamino)-3-oxopropionate 98.0+%, TCI America™
CAS: 29689-63-2 Molecular Formula: C12H15NO3 Molecular Weight (g/mol): 221.26 MDL Number: MFCD28128094 InChI Key: MPAGGJGTAABULX-UHFFFAOYSA-N Synonym: 3-(Benzylamino)-3-oxopropionic Acid Ethyl Ester, Ethyl (N-Benzylcarbamoyl)acetate, (N-Benzylcarbamoyl)acetic Acid Ethyl Ester, Ethyl N-Benzylmalonamate, N-Benzylmalonamic Acid Ethyl Ester PubChem CID: 11379032 IUPAC Name: ethyl 2-(benzylcarbamoyl)acetate SMILES: CCOC(=O)CC(=O)NCC1=CC=CC=C1