missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-benzoylacrylate, predominantly trans, 94%

Catalog No. AAA1942609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1942609 10 g
AAA1942618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1942609 Supplier Thermo Scientific Chemicals Supplier No. A1942609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17450-56-5 | C12H12O3 | 204.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17450-56-5
Molecular Formula C12H12O3
Molecular Weight (g/mol) 204.225
MDL Number MFCD00011533
InChI Key ACXLBHHUHSJENU-CMDGGOBGSA-N
Synonym ethyl 3-benzoylacrylate, ethyl 4-oxo-4-phenylbut-2-enoate, ethyl-3-benzoylacrylate, e-ethyl 4-oxo-4-phenylbut-2-enoate, ethyl trans-3-benzoylacrylate, trans-3-benzoylacrylic acid ethyl ester, ethyl 2e-4-oxo-4-phenylbut-2-enoate, ethyl e-4-oxo-4-phenylbut-2-enoate, trans-3-benzoylacrilic acid ethyl ester, ethyl=3-benzoylacrylate
PubChem CID 5369605
IUPAC Name ethyl (E)-4-oxo-4-phenylbut-2-enoate
SMILES CCOC(=O)C=CC(=O)C1=CC=CC=C1

Specifications

Density 1.12
Boiling Point 184°C to 185°C (25 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.543
Quantity 10 g
UN Number UN2810
Beilstein 2210035
Formula Weight 204.23
Percent Purity 94%
Chemical Name or Material Ethyl 3-benzoylacrylate, predominantly trans
Hazard Category H302-H311-H315-H317-H318-H335
Hazard Statement GHS H Statement
H311-H318-H302-H315-H317-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 408-040-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only