missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminoisonicotinate 98.0+%, TCI America™

Catalog No. E11371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11371G 1 g
E11375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11371G Supplier TCI America Supplier No. E11371G

Chemical Identifiers

CAS 14208-83-4
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD03788270
InChI Key BIFLTHWDFNLOTD-UHFFFAOYSA-N
PubChem CID 2736365
IUPAC Name ethyl 3-aminopyridine-4-carboxylate
SMILES CCOC(=O)C1=C(N)C=NC=C1

Specifications

Melting Point 67°C
Color Yellow
Quantity 1 g
Synonym ethyl 3-aminoisonicotinate, ethyl 3-amino-4-pyridinecarboxylate, 3-amino-isonicotinic acid ethyl ester, 3-aminoisonicotinic acid ethyl ester, 3-amino-4-pyridinecarboxylic acid ethyl ester, 3-aminopyridine-4-carboxylic acid ethyl ester, 4-pyridinecarboxylic acid, 3-amino-, ethyl ester, ethyl3-aminoisonicotinate, pubchem16750, ethyl 3-amino isonicotinate
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Aminoisonicotinate