accessibility menu, dialog, popup

UserName

CAS RN 14208-83-4

H3COONNH2

CAS RN 14208-83-4

IUPAC Name: ethyl 3-aminopyridine-4-carboxylate
Synonyms: ethyl 3-aminopyridine-4-carboxylate
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: BIFLTHWDFNLOTD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)C=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results