Learn More
Ethyl 3-aminocrotonate, 98+%
CAS: 626-34-6 | C6H11NO2 | 129.16 g/mol
$356.59 - $356.59
Chemical Identifiers
| CAS | 626-34-6 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.16 |
| MDL Number | MFCD00008073,MFCD02730138 |
| InChI Key | YPMPTULBFPFSEQ-UHFFFAOYSA-N |
| Synonym | z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate |
| PubChem CID | 643756 |
| IUPAC Name | ethyl (Z)-3-aminobut-2-enoate |
| SMILES | CCOC(=O)C=C(C)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1299136
|
Thermo Scientific Chemicals
A1299136 |
500 g |
Each for $356.59
|
|
|||||
Description
Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.
Solubility
Soluble in water (26 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acids, bases, oxidizing agents, reducing agents. Air & Moisture Sensitive & Hygroscopic.
Chemical Identifiers
| 626-34-6 | |
| 129.16 | |
| YPMPTULBFPFSEQ-UHFFFAOYSA-N | |
| 643756 | |
| CCOC(=O)C=C(C)N |
| C6H11NO2 | |
| MFCD00008073,MFCD02730138 | |
| z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate | |
| ethyl (Z)-3-aminobut-2-enoate |
Specifications
| 626-34-6 | |
| 1.022 | |
| 97°C (206°F) | |
| 1.499 | |
| 500 g | |
| 471284 | |
| z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate | |
| YPMPTULBFPFSEQ-UHFFFAOYSA-N | |
| ethyl (Z)-3-aminobut-2-enoate | |
| 643756 | |
| ≥98% |
| 29°C to 35°C | |
| 210°C to 215°C | |
| C6H11NO2 | |
| MFCD00008073,MFCD02730138 | |
| UN3263 | |
| Air and moisture sensitive; Hygroscopic | |
| Soluble in water (26g/L) (25°C). | |
| CCOC(=O)C=C(C)N | |
| 129.16 | |
| 129.16 | |
| Ethyl 3-aminocrotonate |
Safety and Handling
GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 230-782-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only