missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98+%

Catalog No. aaa1299136
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1299136 Supplier Thermo Scientific Chemicals Supplier No. A1299136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-34-6 | C6H11NO2 | 129.16 g/mol

Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

Solubility
Soluble in water (26 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acids, bases, oxidizing agents, reducing agents. Air & Moisture Sensitive & Hygroscopic.

Chemical Identifiers

CAS 626-34-6
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00008073,MFCD02730138
InChI Key YPMPTULBFPFSEQ-UHFFFAOYSA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Melting Point 29°C to 35°C
Density 1.022
Boiling Point 210°C to 215°C
Flash Point 97°C (206°F)
Refractive Index 1.499
Quantity 500 g
UN Number UN3263
Beilstein 471284
Sensitivity Air and moisture sensitive; Hygroscopic
Solubility Information Soluble in water (26g/L) (25°C).
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-aminocrotonate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 230-782-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only