missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%

Catalog No. p-3736421
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118000100 10 g Glass bottle
AC118000500 50 g Plastic bottle
AC118002500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118000100 Supplier Thermo Scientific Chemicals Supplier No. 118000100
Only null left

CAS: 886-86-2 | C9H11NO2·CH4O3S | 261.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 886-86-2
Molecular Formula C9H11NO2·CH4O3S
Molecular Weight (g/mol) 261.29
MDL Number MFCD00013176
InChI Key FQZJYWMRQDKBQN-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt
PubChem CID 261501
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
SMILES CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O

Specifications

Melting Point 146.0°C to 150.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4CO2C2H5·CH3SO3H
Quantity 10 g
Beilstein 14, 389
Merck Index 15, 9786
Solubility Information (5% in Water) Clear solution
Formula Weight 261.29
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Ethyl 3-aminobenzoate, methanesulfonic acid salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 212-956-8

RUO – Research Use Only