missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Amino-3-methylbutyrate Hydrochloride 98.0+%, TCI America™

Catalog No. E08091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08091G 1 g
E08095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08091G Supplier TCI America Supplier No. E08091G

Chemical Identifiers

CAS 85532-40-7
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
InChI Key HFBJSIGXRKAYAF-UHFFFAOYSA-N
Synonym 3-Amino-3-methylbutyric Acid Ethyl Ester Hydrochloride
PubChem CID 15578558
IUPAC Name ethyl 3-amino-3-methylbutanoate;hydrochloride
SMILES CCOC(=O)CC(C)(C)N.Cl

Specifications

Melting Point 101°C
Color White
Quantity 1 g
Formula Weight 181.66
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Amino-3-methylbutyrate Hydrochloride