missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Acetoxyhexanoate 98.0+%, TCI America™

Catalog No. E10145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10145G 5 g
E101425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10145G Supplier TCI America Supplier No. E10145G
Only null left

Chemical Identifiers

CAS 21188-61-4
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00792508
InChI Key UYBMIHNGXIOMAW-UHFFFAOYSA-N
Synonym 3-Acetoxyhexanoic Acid Ethyl Ester
PubChem CID 89464
IUPAC Name ethyl 3-acetyloxyhexanoate
SMILES CCCC(CC(=O)OCC)OC(=O)C

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 g
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Acetoxyhexanoate