accessibility menu, dialog, popup

UserName

CAS RN 21188-61-4

CH3OOH3COH3CO

CAS RN 21188-61-4

IUPAC Name: ethyl 3-(acetyloxy)hexanoate
Synonyms: ethyl 3-(acetyloxy)hexanoate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: UYBMIHNGXIOMAW-UHFFFAOYNA-N
SMILES: CCCC(CC(=O)OCC)OC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results