missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-methoxybenzoyl)propionate, 98%

Catalog No. AAL0798914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0798914 25 g
AAL0798906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0798914 Supplier Thermo Scientific Chemicals Supplier No. L0798914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15118-67-9 | C13H16O4 | 236.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15118-67-9
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.267
MDL Number MFCD00051784
InChI Key FYUAOZFEVHSJTO-UHFFFAOYSA-N
Synonym ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester
PubChem CID 585132
IUPAC Name ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC

Specifications

Melting Point 49°C to 53°C
Quantity 25 g
Beilstein 2120509
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material Ethyl 3-(4-methoxybenzoyl)propionate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only