CAS RN 15118-67-9
IUPAC Name:
ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Synonyms:
ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Molecular Weight (g/mol):
236.27
Molecular Formula:
C13H16O4
InChi Key:
FYUAOZFEVHSJTO-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCC(=O)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-(4-methoxybenzoyl)propionate, 98%
Ethyl 3-(4-methoxybenzoyl)propionate, 98%
CAS: 15118-67-9 Molecular Formula: C13H16O4 Molecular Weight (g/mol): 236.267 MDL Number: MFCD00051784 InChI Key: FYUAOZFEVHSJTO-UHFFFAOYSA-N Synonym: ethyl 3-4-methoxybenzoyl propionate,ethyl 4-4-methoxyphenyl-4-oxobutanoate,ethyl 4-4-methoxyphenyl-4-oxobutyrate,acmc-1ciqg,ethyl 3-4-methoxybenzoylpropionate,ethyl 3-4-methoxybenzoyl propanoate,ethyl 3-4-methoxybenzoyl-propionate,ethyl 4-4-methoxyphenyl-4-oxobutanoate #,gamma-oxo-4-methoxybenzenebutyric acid ethyl ester,4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester PubChem CID: 585132 IUPAC Name: ethyl 4-(4-methoxyphenyl)-4-oxobutanoate SMILES: CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC