missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate 98.0+%, TCI America™
$255.38 - $891.02
Chemical Identifiers
| CAS | 49744-93-6 |
|---|---|
| Molecular Formula | C12H11NO3 |
| Molecular Weight (g/mol) | 217.224 |
| MDL Number | MFCD00029621 |
| InChI Key | ITKQVWKZLCNILF-UHFFFAOYSA-N |
| Synonym | 3-(4-Cyanophenyl)-3-oxopropionic Acid Ethyl Ester |
| PubChem CID | 2757969 |
| IUPAC Name | ethyl 3-(4-cyanophenyl)-3-oxopropanoate |
| SMILES | CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E1080200MG
|
TCI America
E1080200MG |
200 mg |
Each for $255.38
|
|
|||||
|
E10801G
|
TCI America
E10801G |
1 g |
Each for $891.02
|
|
|||||
Chemical Identifiers
| 49744-93-6 | |
| 217.224 | |
| ITKQVWKZLCNILF-UHFFFAOYSA-N | |
| 2757969 | |
| CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N |
| C12H11NO3 | |
| MFCD00029621 | |
| 3-(4-Cyanophenyl)-3-oxopropionic Acid Ethyl Ester | |
| ethyl 3-(4-cyanophenyl)-3-oxopropanoate |
Specifications
| 49744-93-6 | |
| 140°C | |
| MFCD00029621 | |
| 3-(4-Cyanophenyl)-3-oxopropionic Acid Ethyl Ester | |
| CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N | |
| 217.224 | |
| 217.22 | |
| Crystalline Powder |
| 67°C | |
| C12H11NO3 | |
| 200 mg | |
| ITKQVWKZLCNILF-UHFFFAOYSA-N | |
| ethyl 3-(4-cyanophenyl)-3-oxopropanoate | |
| 2757969 | |
| ≥98.0% (GC,T) | |
| Ethyl 3-(4-Cyanophenyl)-3-oxopropionate |
Safety and Handling
TSCA : No