accessibility menu, dialog, popup

UserName

H3COOON

CAS RN 49744-93-6

IUPAC Name: ethyl 3-(4-cyanophenyl)-3-oxopropanoate
Synonyms: ethyl 3-(4-cyanophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: ITKQVWKZLCNILF-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results