missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 67°C |
| Boiling Point | 140°C |
| Quantity | 200 mg |
| Formula Weight | 217.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 3-(4-Cyanophenyl)-3-oxopropionate |
Safety and Handling
| TSCA | No |