missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-Diethoxyacrylate 95.0+%, TCI America™

Catalog No. p-7127062
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20065G 5 g
D200625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20065G Supplier TCI America Supplier No. D20065G

Chemical Identifiers

CAS 32002-24-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00012220
InChI Key POJGRKZMYVJCST-UHFFFAOYSA-N
Synonym 3,3-Diethoxyacrylic Acid Ethyl Ester
PubChem CID 169388
IUPAC Name ethyl 3,3-diethoxyprop-2-enoate
SMILES CCOC(=CC(=O)OCC)OCC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3,3-Diethoxyacrylate