accessibility menu, dialog, popup

UserName

CAS RN 32002-24-7

IUPAC Name: ethyl 3,3-diethoxyprop-2-enoate
Synonyms: ethyl 3,3-diethoxyprop-2-enoate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: POJGRKZMYVJCST-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results