missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(3,4-Dichlorophenyl)carbazate 98.0+%, TCI America™
$147.25 - $587.17
Chemical Identifiers
| CAS | 13124-15-7 |
|---|---|
| Molecular Formula | C9H10Cl2N2O2 |
| Molecular Weight (g/mol) | 249.09 |
| MDL Number | MFCD28975114 |
| InChI Key | JLVAIPOSBJQZGC-UHFFFAOYSA-N |
| Synonym | 3-(3,4-Dichlorophenyl)carbazic Acid Ethyl Ester, 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine |
| PubChem CID | 12787057 |
| IUPAC Name | N'-(3,4-dichlorophenyl)ethoxycarbohydrazide |
| SMILES | CCOC(=O)NNC1=CC(Cl)=C(Cl)C=C1 |
Chemical Identifiers
| 13124-15-7 | |
| 249.09 | |
| JLVAIPOSBJQZGC-UHFFFAOYSA-N | |
| 12787057 | |
| CCOC(=O)NNC1=CC(Cl)=C(Cl)C=C1 |
| C9H10Cl2N2O2 | |
| MFCD28975114 | |
| 3-(3,4-Dichlorophenyl)carbazic Acid Ethyl Ester, 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine | |
| N'-(3,4-dichlorophenyl)ethoxycarbohydrazide |
Specifications
| 13124-15-7 | |
| White-Yellow | |
| MFCD28975114 | |
| 3-(3,4-Dichlorophenyl)carbazic Acid Ethyl Ester, 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine | |
| CCOC(=O)NNC1=CC(Cl)=C(Cl)C=C1 | |
| 249.09 | |
| 249.09 | |
| Crystalline Powder |
| 112°C | |
| C9H10Cl2N2O2 | |
| 1 g | |
| JLVAIPOSBJQZGC-UHFFFAOYSA-N | |
| N'-(3,4-dichlorophenyl)ethoxycarbohydrazide | |
| 12787057 | |
| ≥98.0% (GC) | |
| Ethyl 3-(3,4-Dichlorophenyl)carbazate |
Safety and Handling
TSCA : No